Assay Details
| Gene Symbol: TNFAIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030726.1 | NP_001025897.1 | 3 - 4 | 734 | 75 | |
| XM_015295802.2 | XP_015151288.2 | 3 - 4 | 550 | 75 | ||
| XM_015295804.2 | XP_015151290.2 | 2 - 3 | 522 | 75 | ||
| GenBank mRNA | AJ851702.1 | - | 3 - 4 | 734 | 75 | |
More Information
Set Membership: |
|

TNFAIP1
Genomic Map