Assay Details
| Gene Symbol: CHPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030764.1 | NP_001025935.1 | 1 - 2 | 314 | 69 | |
| XM_015285482.2 | XP_015140968.1 | 1 - 2 | 318 | 69 | ||
| XM_015285485.2 | XP_015140971.1 | 1 - 2 | 308 | 69 | ||
| XM_015285488.2 | XP_015140974.1 | 1 - 2 | 619 | 69 | ||
| GenBank mRNA | AJ721079.1 | - | 1 - 2 | 314 | 69 | |
More Information
Set Membership: |
|

CHPT1
Genomic Map