Assay Details
| Gene Symbol: CHRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030765.1 | NP_001025936.1 | 1 | 1153 | 92 | |
| XM_015285631.2 | XP_015141117.1 | 2 | 1331 | 92 | ||
| XM_015285640.2 | XP_015141126.1 | 3 | 1397 | 92 | ||
More Information
Set Membership: |
|

CHRM2
Genomic Map