Assay Details
| Gene Symbol: CDPF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030772.1 | NP_001025943.1 | 2 - 3 | 343 | 108 | |
| GenBank mRNA | AJ720166.1 | - | 2 - 3 | 343 | 108 | |
More Information
Set Membership: |
|

CDPF1
Genomic Map