Assay Details
| Gene Symbol: RCSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030789.1 | NP_001025960.1 | 6 - 7 | 1281 | 103 | |
| XM_015296322.2 | XP_015151808.2 | 7 - 8 | 1383 | 103 | ||
| XM_015296326.2 | XP_015151812.2 | 6 - 7 | 1151 | 103 | ||
| XM_015296328.2 | XP_015151814.2 | 5 - 6 | 1239 | 103 | ||
| XM_025149388.1 | XP_025005156.1 | 6 - 7 | 1294 | 103 | ||
| XM_025149389.1 | XP_025005157.1 | 6 - 7 | 1180 | 103 | ||
| GenBank mRNA | AJ719861.1 | - | 6 - 7 | 1281 | 103 | |
More Information
Set Membership: |
|

RCSD1
Genomic Map