Assay Details
| Gene Symbol: USP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031290.1 | NP_001026461.1 | 8 - 9 | 1801 | 81 | |
| XM_015291024.2 | XP_015146510.1 | 7 - 8 | 1626 | 81 | ||
| GenBank mRNA | AJ720494.1 | - | 8 - 9 | 1801 | 81 | |
More Information
Set Membership: |
|

USP1
Genomic Map