Assay Details
| Gene Symbol: CERK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031340.1 | NP_001026511.1 | 2 - 3 | 381 | 72 | |
| XM_015275753.2 | XP_015131239.1 | 2 - 3 | 248 | 72 | ||
| GenBank mRNA | AJ851677.1 | - | 2 - 3 | 381 | 72 | |
| CR386590.1 | - | 1 - 2 | 49 | 72 | ||
More Information
Set Membership: |
|

CERK
Genomic Map