Assay Details
| Gene Symbol: MANSC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031381.2 | NP_001026552.1 | 2 - 3 | 401 | 116 | |
| GenBank mRNA | AJ719946.1 | - | 2 - 3 | 402 | 116 | |
More Information
Set Membership: |
|

MANSC1
Genomic Map