Assay Details
| Gene Symbol: IDH3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031387.1 | NP_001026558.1 | 7 - 8 | 677 | 65 | |
| GenBank mRNA | AJ720045.1 | - | 7 - 8 | 677 | 65 | |
More Information
Set Membership: |
|

IDH3B
Genomic Map