Assay Details
| Gene Symbol: DNASE1L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034815.1 | NP_001029987.1 | 1 - 2 | 127 | 72 | |
| GenBank mRNA | BX929513.2 | - | 1 - 2 | 106 | 72 | |
| DQ151449.1 | - | 1 - 2 | 127 | 72 | ||
More Information
Set Membership: |
|

DNASE1L3
Genomic Map