Assay Details
| Gene Symbol: MORF4L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037173.1 | NP_001032250.1 | 4 - 5 | 263 | 87 | |
| NM_001037176.1 | NP_001032253.1 | 4 - 5 | 263 | 87 | ||
| XM_015292126.2 | XP_015147612.1 | 5 - 6 | 343 | 87 | ||
| XM_015292127.2 | XP_015147613.1 | 5 - 6 | 372 | 87 | ||
| XM_015292128.2 | XP_015147614.1 | 5 - 6 | 412 | 87 | ||
| GenBank mRNA | BI389851.1 | - | 4 - 5 | 255 | 87 | |
| BI391102.1 | - | 4 - 5 | 263 | 87 | ||
| BX930523.1 | - | 4 - 5 | 255 | 87 | ||
| CR386790.1 | - | 4 - 5 | 227 | 87 | ||
More Information
Set Membership: |
|

MORF4L2
Genomic Map