Assay Details
| Gene Symbol: WNT5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037269.1 | NP_001032346.1 | 3 - 4 | 624 | 58 | |
| XM_015285801.2 | XP_015141287.1 | 4 - 5 | 1004 | 58 | ||
| GenBank mRNA | AY753289.1 | - | 3 - 4 | 624 | 58 | |
More Information
Set Membership: |
|

WNT5B
Genomic Map