Assay Details
| Gene Symbol: CDK2AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039264.3 | NP_001034353.3 | 3 - 4 | 575 | 75 | |
| XM_015275271.2 | XP_015130757.1 | 4 - 5 | 452 | 75 | ||
| XM_015275272.2 | XP_015130758.1 | 3 - 4 | 342 | 75 | ||
| XM_015275273.2 | XP_015130759.1 | 3 - 4 | 385 | 75 | ||
| XM_015275274.2 | XP_015130760.1 | 3 - 4 | 528 | 75 | ||
| XM_025155452.1 | XP_025011220.1 | 3 - 4 | 339 | 75 | ||
| GenBank mRNA | AJ719364.1 | - | 3 - 4 | 363 | 75 | |
More Information
Set Membership: |
|

CDK2AP1
Genomic Map