Assay Details
| Gene Symbol: CORO1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277868.2 | NP_001264797.2 | 2 - 3 | 601 | 58 | |
| NM_001277869.2 | NP_001264798.1 | 2 - 3 | 328 | 58 | ||
| XM_025155467.1 | XP_025011235.1 | 2 - 3 | 314 | 58 | ||
| XM_025155468.1 | XP_025011236.1 | 2 - 3 | 365 | 58 | ||
| XM_025155469.1 | XP_025011237.1 | 2 - 3 | 229 | 58 | ||
| GenBank mRNA | AJ449366.1 | - | 2 - 3 | 297 | 58 | |
| AJ453793.1 | - | 2 - 3 | 328 | 58 | ||
| AJ719514.1 | - | 2 - 3 | 292 | 58 | ||
| AJ719663.1 | - | 2 - 3 | 290 | 58 | ||
| AJ720924.1 | - | 927 | 58 | |||
| BX930410.1 | - | 2 - 3 | 601 | 58 | ||
| BX932518.2 | - | 2 - 3 | 601 | 58 | ||
| CN383670.1 | - | 2 - 3 | 292 | 58 | ||
More Information
Set Membership: |
|

CORO1C
Genomic Map