Assay Details
| Gene Symbol: FOXN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039284.2 | NP_001034373.2 | 1 - 2 | 556 | 95 | |
| XM_015278070.2 | XP_015133556.1 | 2 - 3 | 598 | 95 | ||
| XM_015278071.2 | XP_015133557.1 | 3 - 4 | 1009 | 95 | ||
| XM_015278072.2 | XP_015133558.1 | 3 - 4 | 5302 | 95 | ||
| XM_015278073.2 | XP_015133559.1 | 2 - 3 | 695 | 95 | ||
| XM_015278074.2 | XP_015133560.1 | 2 - 3 | 601 | 95 | ||
| GenBank mRNA | AJ721059.1 | - | 2 - 3 | 97 | ||
More Information
Set Membership: |
|

FOXN2
Genomic Map