Assay Details
| Gene Symbol: FOXN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039284.2 | NP_001034373.2 | 4 - 5 | 808 | 98 | |
| XM_015278070.2 | XP_015133556.1 | 5 - 6 | 850 | 98 | ||
| XM_015278071.2 | XP_015133557.1 | 6 - 7 | 1261 | 98 | ||
| XM_015278072.2 | XP_015133558.1 | 6 - 7 | 5554 | 98 | ||
| XM_015278073.2 | XP_015133559.1 | 5 - 6 | 947 | 98 | ||
| XM_015278074.2 | XP_015133560.1 | 5 - 6 | 850 | 98 | ||
| GenBank mRNA | AJ721059.1 | - | 5 - 6 | 851 | 98 | |
More Information
Set Membership: |
|

FOXN2
Genomic Map