Assay Details
| Gene Symbol: FDFT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039294.1 | NP_001034383.1 | 2 - 3 | 281 | 74 | |
| GenBank mRNA | AJ719973.1 | - | 2 - 3 | 281 | 74 | |
More Information
Set Membership: |
|

FDFT1
Genomic Map