Assay Details
| Gene Symbol: ZFYVE27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039304.1 | NP_001034393.1 | 5 - 6 | 698 | 56 | |
| XM_015288631.2 | XP_015144117.1 | 5 - 6 | 696 | 56 | ||
| GenBank mRNA | AJ719898.1 | - | 5 - 6 | 698 | 56 | |
More Information
Set Membership: |
|

ZFYVE27
Genomic Map