Assay Details
| Gene Symbol: PRDX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039329.2 | NP_001034418.1 | 1 - 2 | 111 | 54 | |
| GenBank mRNA | AJ720480.1 | - | 1 - 2 | 113 | 54 | |
| BX931632.1 | - | 1 - 2 | 141 | 54 | ||
More Information
Set Membership: |
|

PRDX6
Genomic Map