Assay Details
| Gene Symbol: PRDX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039329.2 | NP_001034418.1 | 4 - 5 | 561 | 75 | |
| GenBank mRNA | AJ720480.1 | - | 4 - 5 | 563 | 75 | |
More Information
Set Membership: |
|

PRDX6
Genomic Map