Assay Details
| Gene Symbol: VPS4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006378.2 | NP_001006378.2 | 6 - 7 | 740 | 71 | |
| GenBank mRNA | AJ719395.1 | - | 6 - 7 | 740 | 71 | |
More Information
Set Membership: |
|

VPS4B
Genomic Map