Assay Details
| Gene Symbol: SELENOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006557.4 | NP_001006557.3 | 3 - 4 | 456 | 95 | |
| GenBank mRNA | AJ450766.1 | - | 3 - 4 | 459 | 95 | |
| AJ720396.1 | - | 3 - 4 | 392 | 95 | ||
| BM439905.1 | - | 3 - 4 | 416 | 95 | ||
| CN224003.1 | - | 3 - 4 | 415 | 95 | ||
| DR419695.1 | - | 3 - 4 | 450 | 95 | ||
More Information
Set Membership: |
|

SELENOT
Genomic Map