Assay Details
| Gene Symbol: SELENOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006557.4 | NP_001006557.3 | 5 - 6 | 705 | 57 | |
| GenBank mRNA | AJ720396.1 | - | 5 - 6 | 641 | 57 | |
| CV862139.1 | - | 1 - 2 | 181 | 57 | ||
| DR419695.1 | - | 5 - 6 | 699 | 57 | ||
More Information
Set Membership: |
|

SELENOT
Genomic Map