Assay Details
| Gene Symbol: GUCA1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007881.2 | NP_001007882.1 | 526 | 66 | ||
| GenBank mRNA | S82200.1 | - | 526 | 66 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GUCA1B| Gene Symbol: GUCA1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007881.2 | NP_001007882.1 | 526 | 66 | ||
| GenBank mRNA | S82200.1 | - | 526 | 66 | ||
Set Membership: |
|