Assay Details
Gene Symbol: CELA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032390.2 | NP_001027562.2 | 3 - 4 | 251 | 98 | |
GenBank mRNA | BX931608.2 | - | 3 - 4 | 235 | 98 | |
BX933641.1 | - | 3 - 4 | 226 | 98 | ||
BX933648.1 | - | 3 - 4 | 233 | 98 | ||
BX933649.2 | - | 3 - 4 | 233 | 98 | ||
BX933653.2 | - | 3 - 4 | 248 | 98 | ||
BX933662.1 | - | 3 - 4 | 216 | 98 | ||
BX933671.1 | - | 3 - 4 | 226 | 98 | ||
BX933680.2 | - | 3 - 4 | 225 | 98 | ||
BX933686.1 | - | 3 - 4 | 251 | 98 | ||
BX933689.1 | - | 3 - 4 | 245 | 98 | ||
BX935221.1 | - | 3 - 4 | 221 | 98 | ||
BX935254.1 | - | 3 - 4 | 261 | 98 | ||
BX935312.1 | - | 3 - 4 | 223 | 98 | ||
BX935323.2 | - | 3 - 4 | 233 | 98 | ||
BX935324.2 | - | 3 - 4 | 225 | 98 | ||
BX935330.2 | - | 3 - 4 | 213 | 98 | ||
BX936261.2 | - | 3 - 4 | 252 | 98 | ||
CR338993.2 | - | 3 - 4 | 233 | 98 | ||
CR338996.1 | - | 3 - 4 | 235 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
