Assay Details
| Gene Symbol: ASIC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040467.1 | NP_001035557.1 | 3 - 4 | 710 | 71 | |
| XM_015300232.2 | XP_015155718.1 | 2 - 3 | 3184 | 71 | ||
| XM_015300233.2 | XP_015155719.1 | 2 - 3 | 1693 | 71 | ||
| XM_015300234.2 | XP_015155720.1 | 4 - 5 | 1184 | 71 | ||
| XM_015300235.2 | XP_015155721.1 | 4 - 5 | 869 | 71 | ||
| GenBank mRNA | AY956393.1 | - | 3 - 4 | 710 | 71 | |
More Information
Set Membership: |
|

ASIC1
Genomic Map