Assay Details
| Gene Symbol: FGF16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044650.1 | NP_001038115.1 | 1 - 2 | 272 | 96 | |
| GenBank mRNA | DQ640740.1 | - | 1 - 2 | 272 | 96 | |
More Information
Set Membership: |
|

FGF16
Genomic Map