Assay Details
| Gene Symbol: OPN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044653.1 | NP_001038118.1 | 4 - 5 | 783 | 72 | |
| XM_015287980.1 | XP_015143466.1 | 5 - 6 | 930 | 72 | ||
| GenBank mRNA | AB255031.1 | - | 4 - 5 | 521 | 72 | |
| AB295599.1 | - | 4 - 5 | 521 | 72 | ||
| AY882944.1 | - | 4 - 5 | 783 | 72 | ||
| EU124632.1 | - | 4 - 5 | 541 | 72 | ||
| EU124633.1 | - | 4 - 5 | 541 | 72 | ||
| EU124634.1 | - | 4 - 5 | 541 | 72 | ||
More Information
Set Membership: |
|

OPN4
Genomic Map