Assay Details
| Gene Symbol: LGI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044655.2 | NP_001038120.1 | 6 - 7 | 829 | 83 | |
| XM_015288602.2 | XP_015144088.1 | 7 - 8 | 949 | 83 | ||
| XM_015288603.2 | XP_015144089.1 | 6 - 7 | 940 | 83 | ||
| XM_015288606.2 | XP_015144092.1 | 5 - 6 | 3319 | 83 | ||
| XM_015288607.2 | XP_015144093.1 | 7 - 8 | 981 | 83 | ||
| XM_025151593.1 | XP_025007361.1 | 6 - 7 | 877 | 83 | ||
| XM_025151594.1 | XP_025007362.1 | 5 - 6 | 801 | 83 | ||
| XM_025151595.1 | XP_025007363.1 | 6 - 7 | 886 | 83 | ||
| XM_025151596.1 | XP_025007364.1 | 5 - 6 | 868 | 83 | ||
| GenBank mRNA | AY617122.1 | - | 6 - 7 | 830 | 83 | |
| BX933910.2 | - | 6 - 7 | 881 | 83 | ||
More Information
Set Membership: |
|

LGI1
Genomic Map