Assay Details
| Gene Symbol: COLEC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044665.1 | NP_001038130.1 | 2 - 3 | 208 | 71 | |
| XM_015284842.2 | XP_015140328.1 | 3 - 4 | 8919 | 71 | ||
| XM_015284843.2 | XP_015140329.1 | 3 - 4 | 431 | 71 | ||
| XM_015284844.2 | XP_015140330.1 | 2 - 3 | 323 | 71 | ||
| XM_025148811.1 | XP_025004579.1 | 2 - 3 | 122 | 71 | ||
| GenBank mRNA | DQ129669.1 | - | 2 - 3 | 208 | 71 | |
More Information
Set Membership: |
|

COLEC11
Genomic Map