Assay Details
| Gene Symbol: CYP51A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048077.1 | NP_001041542.1 | 5 - 6 | 545 | 68 | |
| XM_015281391.2 | XP_015136877.1 | 3 - 4 | 496 | 68 | ||
| XM_015281392.2 | XP_015136878.1 | 7 - 8 | 2817 | 68 | ||
| GenBank mRNA | AB243875.1 | - | 5 - 6 | 545 | 68 | |
| AB243876.1 | - | 3 - 4 | 358 | 68 | ||
More Information
Set Membership: |
|

CYP51A1
Genomic Map