Assay Details
| Gene Symbol: AP2M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079494.1 | NP_001072962.1 | 6 - 7 | 808 | 68 | |
| XM_015291442.2 | XP_015146928.1 | 8 - 9 | 798 | 68 | ||
| XM_015291443.2 | XP_015146929.1 | 8 - 9 | 868 | 68 | ||
| XM_015291444.2 | XP_015146930.1 | 7 - 8 | 807 | 68 | ||
| XM_015291445.2 | XP_015146931.1 | 7 - 8 | 815 | 68 | ||
| XM_015291446.2 | XP_015146932.1 | 7 - 8 | 879 | 68 | ||
| XM_015291447.2 | XP_015146933.1 | 6 - 7 | 793 | 68 | ||
| XM_025153417.1 | XP_025009185.1 | 6 - 7 | 823 | 68 | ||
| GenBank mRNA | AJ719338.1 | - | 6 - 7 | 808 | 68 | |
More Information
Set Membership: |
|

AP2M1
Genomic Map