Assay Details
| Gene Symbol: CRLF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079719.3 | NP_001073187.2 | 5 - 6 | 840 | 54 | |
| GenBank mRNA | AJ720057.1 | - | 4 - 5 | 627 | 54 | |
| BU290806.1 | - | 1 - 2 | 190 | 54 | ||
More Information
Set Membership: |
|

CRLF3
Genomic Map