Assay Details
| Gene Symbol: MINA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079723.1 | NP_001073191.1 | 9 - 10 | 1296 | 57 | |
| GenBank mRNA | AJ720509.1 | - | 9 - 10 | 1296 | 57 | |
More Information
Set Membership: |
|

MINA
Genomic Map