Assay Details
| Gene Symbol: SHOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079724.1 | NP_001073192.1 | 3 - 4 | 572 | 69 | |
| GenBank mRNA | DQ675517.1 | - | 3 - 4 | 572 | 69 | |
More Information
Set Membership: |
|

SHOX
Genomic Map