Assay Details
| Gene Symbol: RCOR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079727.1 | NP_001073195.1 | 8 - 9 | 991 | 64 | |
| XM_015283668.2 | XP_015139154.2 | 9 - 10 | 1088 | 64 | ||
| XM_015283669.2 | XP_015139155.1 | 9 - 10 | 991 | 64 | ||
| XM_015283670.2 | XP_015139156.1 | 9 - 10 | 1372 | 64 | ||
| XM_015283671.2 | XP_015139157.2 | 9 - 10 | 1070 | 64 | ||
| XM_015283672.2 | XP_015139158.1 | 10 - 11 | 1348 | 64 | ||
| XM_015283677.2 | XP_015139163.2 | 9 - 10 | 1088 | 64 | ||
| XM_015283678.2 | XP_015139164.2 | 9 - 10 | 1070 | 64 | ||
| XM_025148670.1 | XP_025004438.1 | 6 - 7 | 807 | 64 | ||
| XM_025148671.1 | XP_025004439.1 | 6 - 7 | 802 | 64 | ||
| XM_025148672.1 | XP_025004440.1 | 7 - 8 | 824 | 64 | ||
| GenBank mRNA | AJ720594.1 | - | 8 - 9 | 991 | 64 | |
More Information
Set Membership: |
|

RCOR3
Genomic Map