Assay Details
| Gene Symbol: ZBTB33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079729.1 | NP_001073197.1 | 1 | 1954 | 117 | |
| XM_015278297.2 | XP_015133783.1 | 2 | 2061 | 117 | ||
| XM_015278298.2 | XP_015133784.1 | 2 | 2126 | 117 | ||
| GenBank mRNA | EF028066.1 | - | 1 | 1954 | 117 | |
More Information
Set Membership: |
|

ZBTB33
Genomic Map