Assay Details
| Gene Symbol: TMEM38A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079758.1 | NP_001073226.1 | 3 - 4 | 510 | 78 | |
| XM_025144285.1 | XP_025000053.1 | 4 - 5 | 577 | 78 | ||
| GenBank mRNA | AJ720241.1 | - | 3 - 4 | 510 | 78 | |
More Information
Set Membership: |
|

TMEM38A
Genomic Map