Assay Details
| Gene Symbol: CRYZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079763.1 | NP_001073231.1 | 5 - 6 | 760 | 62 | |
| XM_015291065.2 | XP_015146551.1 | 4 - 5 | 839 | 62 | ||
| XM_025152903.1 | XP_025008671.1 | 5 - 6 | 834 | 62 | ||
| GenBank mRNA | AJ721051.1 | - | 5 - 6 | 760 | 62 | |
More Information
Set Membership: |
|

CRYZ
Genomic Map