Assay Details
| Gene Symbol: PPP6C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079765.1 | NP_001073233.1 | 6 - 7 | 706 | 75 | |
| GenBank mRNA | AJ720684.1 | - | 6 - 7 | 706 | 75 | |
| CR389409.1 | - | 6 - 7 | 681 | 75 | ||
More Information
Set Membership: |
|

PPP6C
Genomic Map