Assay Details
| Gene Symbol: GPR34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080886.1 | NP_001074355.1 | 1 | 949 | 90 | |
| XM_015301571.2 | XP_015157057.1 | 2 | 1056 | 90 | ||
| GenBank mRNA | BX929365.1 | - | 2 | 1043 | 90 | |
More Information
Set Membership: |
|

GPR34
Genomic Map