Assay Details
| Gene Symbol: PTGER4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081503.1 | NP_001074972.1 | 2 - 3 | 1200 | 87 | |
| XM_015277319.2 | XP_015132805.1 | 1 - 2 | 1397 | 87 | ||
| GenBank mRNA | EF200121.1 | - | 2 - 3 | 1200 | 87 | |
More Information
Set Membership: |
|

PTGER4
Genomic Map