Assay Details
| Gene Symbol: TNFSF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083361.1 | NP_001076830.1 | 3 - 4 | 641 | 105 | |
| XM_015275777.2 | XP_015131263.1 | 2 - 3 | 395 | 105 | ||
| GenBank mRNA | EF379383.1 | - | 3 - 4 | 641 | 105 | |
More Information
Set Membership: |
|

TNFSF11
Genomic Map