Assay Details
| Gene Symbol: GTF2H5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097535.1 | NP_001091004.1 | 2 - 3 | 157 | 71 | |
| XM_015284124.2 | XP_015139610.1 | 2 - 3 | 177 | 71 | ||
| XM_015284125.2 | XP_015139611.1 | 2 - 3 | 223 | 71 | ||
| GenBank mRNA | AJ720114.1 | - | 2 - 3 | 157 | 71 | |
More Information
Set Membership: |
|

GTF2H5
Genomic Map