Assay Details
| Gene Symbol: IL17D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045037.1 | 1 | 252 | 67 | ||
| GenBank mRNA | EF570583.1 | - | 1 | 252 | 67 | |
| KT899946.1 | - | 2 | 413 | 67 | ||
More Information
Set Membership: |
|

IL17D
Genomic Map