Assay Details
| Gene Symbol: DTYMK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278350.1 | NP_001265279.1 | 4 - 5 | 574 | 109 | |
| GenBank mRNA | AJ719278.1 | - | 2 - 3 | 1681 | 109 | |
| BU134642.1 | - | 2 - 3 | 258 | 109 | ||
| BU305074.1 | - | 1 - 2 | 97 | 109 | ||
| BU319316.1 | - | 4 - 5 | 512 | 109 | ||
| BX935908.2 | - | 4 - 5 | 574 | 109 | ||
More Information
Set Membership: |
|

DTYMK
Genomic Map