Assay Details
| Gene Symbol: GUCA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204320.2 | NP_989651.1 | 627 | 98 | ||
| GenBank mRNA | S82199.1 | - | 627 | 98 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GUCA1A| Gene Symbol: GUCA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204320.2 | NP_989651.1 | 627 | 98 | ||
| GenBank mRNA | S82199.1 | - | 627 | 98 | ||
Set Membership: |
|