Assay Details
| Gene Symbol: RPL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204581.4 | NP_989912.1 | 6 - 7 | 797 | 71 | |
| GenBank mRNA | U31753.1 | - | 2 - 3 | 192 | 71 | |
| X57016.1 | - | 797 | 71 | |||
More Information
Set Membership: |
|

RPL5
Genomic Map