Assay Details
| Gene Symbol: SRSF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113741.1 | NP_001107213.1 | 2 - 3 | 517 | 66 | |
| GenBank mRNA | AJ719371.1 | - | 2 - 3 | 517 | 66 | |
More Information
Set Membership: |
|

SRSF1
Genomic Map