Assay Details
| Gene Symbol: UGP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204111.2 | NP_989442.2 | 5 - 6 | 703 | 81 | |
| XM_015278016.2 | XP_015133502.1 | 5 - 6 | 596 | 81 | ||
| GenBank mRNA | AF534111.1 | - | 5 - 6 | 689 | 81 | |
| AJ719970.1 | - | 5 - 6 | 703 | 81 | ||
More Information
Set Membership: |
|

UGP2
Genomic Map